missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Anhydrous, TRC

Product Code. 30476435
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
Unit Size:
25g
500g
5g
Product Code. Quantity unitSize
30476435 25 g 25g
30472064 500 g 500g
30471123 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 30476435

Brand: Toronto Research Chemicals TRCM11033325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Formula Weight 93.9227
InChI Formula InChI=1S/2ClH.Mg/h2*1H;/q;;+2/p-2
IUPAC Name magnesium;dichloride
Molecular Formula MgCl2
Molecular Weight (g/mol) 95.21
SMILES [Mg+2].[Cl-].[Cl-]
Synonym Aerotex Accelerator MX, C-TEK, Catalyst G, Ekimac, FIX-Mg, Freecat MX, HMC 23D, Magnesium Dichloride, Magnesium(II) Chloride, Magnogene, NSC 529832, Soft Wafer, TMT 2, TMT 4, White Nigari, White Nigari NS
Chemical Name or Material Magnesium Chloride anhydrous

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.