missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride, Anhydrous, MP Biomedicals

Product Code. 11421856 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421856 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421856 Supplier MP Biomedicals Supplier No. 0520984480
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7786-30-3
Quantity 100 g
Molecular Formula Cl2Mg
MDL Number MFCD00011106
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES [Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) dichloride
Molecular Weight (g/mol) 95.21
PubChem CID 5360315
ChEBI CHEBI:6636
Formula Weight 95.22
Chemical Name or Material Magnesium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.