missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous

Product Code. 10604005 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10604005 50 g Plastic bottle 50g
10031524 10 g Plastic bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10604005 Supplier Thermo Scientific Acros Supplier No. 197060500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
IUPAC Name magnesium(2+) dichloride
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Boiling Point 1412.0°C
Melting Point 708.0°C
Color White
Physical Form Beads
Quantity 50 g
Assay Percent Range 99.9%
Linear Formula MgCl2
Fieser 11,310
Merck Index 15, 5726
Solubility Information Solubility in water: 540g/L (20°C)
Formula Weight 95.21
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Magnesium chloride, <10 mesh, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.