missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride 50% (w/w) Solution, Certified AR, for Analysis, Fisher Chemical

Product Code. 10111770 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10111770 2.5 L Amber glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10111770 Supplier Fisher Chemical Supplier No. M/0602/17
Quantity Price
1 £267.66 / 2.5 L
4 £223.33 / 2.5 L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7786-30-3
Color White
Physical Form Liquid
Quantity 2.5 L
Molecular Formula Cl2Mg
MDL Number MFCD00011106
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES [Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) dichloride
Molecular Weight (g/mol) 95.21
PubChem CID 5360315
ChEBI CHEBI:6636
Formula Weight 95.21g/mol
Packaging Amber glass bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.