missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Magnesium carbonate hydroxide pentahydrate, for biochemistry, 40.0-43.5% Magnesium oxide

Product Code. 12604175 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 12604175

Brand: Thermo Scientific Chemicals 448040025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56378-72-4 | C4H12Mg5O19 | 485.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56378-72-4
Molecular Formula C4H12Mg5O19
Molecular Weight (g/mol) 485.65
MDL Number MFCD00149784
InChI Key SXKPPTNZIUYFJR-UHFFFAOYSA-D
Synonym acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5
IUPAC Name pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 350.0°C
Quantity 2.5 kg
Linear Formula Mg(OH)2·5H2O
Formula Weight 485.69
Percent Purity 40.0 to 43.5% (Magnesium o)
Grade Biochemical
Packaging Plastic Drum
Chemical Name or Material Magnesium carbonate hydroxide pentahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.