missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate hydroxide hydrate, Puratronic™, 99.996% (metals basis)

Product Code. 11346897
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346897 25 g 25g
11336897 5 g 5g
11356897 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346897 Supplier Thermo Scientific Alfa Aesar Supplier No. 010796.14
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol

Magnesium carbonate hydroxide hydrate is used as drier, color ficatives, anticaking agent, carrier, swelling agent, acidity regulator. Magnesium carbonate is the production of magnesium oxide by calcining. Magnesite and dolomite minerals are used to produce refractory bricks. MgCO3 is also used in flooring, fireproofing, fire extinguishing compositions, cosmetics, dusting powder, and toothpaste. Other applications are as filler material, smoke suppressant in plastics, a reinforcing agent in neoprene rubber, a drying agent, a laxative to loosen the bowels, and color retention in foods. In addition, high purity magnesium carbonate is used as antacid and as an additive in table salt to keep it free flowing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
Synonym Magnesium carbonate, basic; Magnesium hydroxide carbonate
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Merck Index 14,5660
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 142.63 (Anhydrous)
Percent Purity 99.996%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Magnesium carbonate hydroxide hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.