missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate basic, Honeywell™

Product Code. 15666060 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15666060 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15666060 Supplier Honeywell Research Chemicals Supplier No. 630621KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum p.a., heavy,≥40% (MgO)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

CAS 39409-82-0
Quantity 1 kg
Molecular Formula C4H10Mg5O18
Linear Formula MgCO3
MDL Number MFCD00149783
UN Number NONH for all modes of transport
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
Molecular Weight (g/mol) 467.63
Formula Weight 84.31g/mol
Percent Purity ≥40% (MgO)
Chemical Name or Material Magnesium carbonate basic
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.