missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide hexahydrate, 99+%, for analysis

Product Code. 10593561 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10593561 1 kg Glass bottle 1kg
10703591 5 g Glass bottle 5g
10318630 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10593561 Supplier Thermo Scientific Acros Supplier No. 202510010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-53-2 | Br2H12MgO6 | 292.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-53-2
Molecular Formula Br2H12MgO6
Molecular Weight (g/mol) 292.20
MDL Number MFCD00149780
InChI Key LGLXXNHIGIJYQQ-UHFFFAOYSA-L
Synonym acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
PubChem CID 17749097
IUPAC Name dibromomagnesium hexahydrate
SMILES O.O.O.O.O.O.Br[Mg]Br

Specifications

Melting Point 165.0°C
Color White
pH 6.5 to 8 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99% min. (Complexometry)
Linear Formula MgBr2·6H2O
Fieser 13,259; 15,194; 16,199
Merck Index 14, 5659
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 292.21
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Magnesium bromide hexahydrate, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium bromide hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.