missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide hexahydrate, 98+%

Product Code. 11428630
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11428630

Brand: Thermo Scientific Alfa Aesar 046120.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-53-2 | Br2H12MgO6 | 292.20 g/mol

Magnesium bromide hexahydrate is used as flame retardant. Used as source of magnesium ion and a co-factor for many enzymes including deoxyribonuclease (DNase) and various restriction enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-53-2
Molecular Formula Br2H12MgO6
Molecular Weight (g/mol) 292.20
MDL Number MFCD00149780
InChI Key LGLXXNHIGIJYQQ-UHFFFAOYSA-L
Synonym acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
PubChem CID 17749097
SMILES O.O.O.O.O.O.Br[Mg]Br

Specifications

Melting Point 172°C
Color Colorless
Physical Form Crystalline
Quantity 500 g
Linear Formula MgBr2·6H2O
Solubility Information Soluble in water, somewhat soluble in alcohol
IUPAC Name magnesium;dibromide;hexahydrate
Formula Weight 292.22 (184.13 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.00 g/mL
Chemical Name or Material Magnesium bromide hexahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.