missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide, 98%, pure, anhydrous

Product Code. 10351290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10351290 1 g Glass bottle 1g
10024157 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351290 Supplier Thermo Scientific Acros Supplier No. 413390010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7789-48-2 | Br2Mg | 184.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-48-2
Molecular Formula Br2Mg
Molecular Weight (g/mol) 184.11
MDL Number MFCD27995637
InChI Key OTCKOJUMXQWKQG-UHFFFAOYSA-L
Synonym magnesium bromide, magnesium dibromide, dibromomagnesium, magnesium-bromide, acmc-20aklq, magnesium bromide salt, magnesium ii bromide, magnesium 2+ ion dibromide, grignard reagent
PubChem CID 82241
IUPAC Name magnesium;dibromide
SMILES [Mg++].[Br-].[Br-]

Specifications

Melting Point 711.0°C
Quantity 1 g
Assay Percent Range 97.5 to 102.5% (Argentometry)
Linear Formula MgBr2
Fieser 02,254; 06,270; 08,273; 10,14; 15,194; 16,199
Merck Index 15,5723
Solubility Information Solubility in water: 1015g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 184.11
Percent Purity 98%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Magnesium bromide, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium bromide, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.