missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium, AZ61A, M11610, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Magnesium, AZ61A, M11610, 50g
Specifications
Specifications
| Chemical Name or Material | Beryllium/Nickel/Lead/Iron/Copper/Tin/Silicon/Manganese/Zinc/Aluminium |
| CAS | 7439-95-4 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Beryllium 0.0018 %/Nickel 0.0025 %/Lead 0.0075 %/Iron 0.008 %/Copper 0.013 %/Tin 0.032 %/Silicon 0.052 %/Manganese 0.067 %/Zinc 1.32 %/Aluminium 5.87 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction