missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium, AS21A, M10210, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Magnesium, AS21A, M10210, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Beryllium/Nickel/Copper/Tin/Lead/Zinc/Aluminium/Iron/Manganese/Silicon |
| CAS | 7439-95-4 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Beryllium 0.0007 %/Nickel 0.0026 %/Copper 0.0045 %/Tin 0.0055 %/Lead 0.006 %/Zinc 0.072 %/Aluminium 2.31 %/Iron (0.004) %/Manganese (0.26) %/Silicon (0.97) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction