missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Aluminum Zinc plate, alloy AZ31B, 6.35mm (0.25in) thick, Thermo Scientific Chemicals

 

Brand:  Thermo Scientific Alfa Aesar 014066.RF

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15406025

  • £258.00 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Magnesium Aluminum Zinc plate, alloy is used in chemical research, structural components and metallurgical coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Magnesium Aluminum Zinc plate, Alloy AZ31B
Mg:Al:Zn; 96:3:1 wt%
30 x 30 cm
Odorless
Plate
6.35mm (0.25 in.) thick
MFCD01074901
Insoluble in water.
≈994g/30x30cm
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only