missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate Tetrahydrate, TRC

Product Code. 30476295
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476295 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476295 Supplier Toronto Research Chemicals Supplier No. TRCM262963100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Acetate Tetrahydrate
CAS 16674-78-5
Molecular Formula C4H14MgO8
Quantity 100 g
Synonym Magnesium acetate tetrahydrate
InChI Formula InChI=1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2
SMILES O.O.O.O.[Mg+2].CC(=O)[O-].CC(=O)[O-]
IUPAC Name magnesium;diacetate;tetrahydrate
Molecular Weight (g/mol) 214.45
Formula Weight 214.0539
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.