Learn More
Magnesium acetate tetrahydrate, Puratronic™, 99.997% (metals basis)
Description
Used as a source of magnesium in biological reactions, and as a source of acetate ion in chemical transformations. Acts as an emission control agent in coal combustion process to reduce acid rain. It is involved in glass formation process via the thermal decomposition of solids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 80°C |
| Color | White |
| Physical Form | Crystals |
| Quantity | 100 g |
| Assay Percent Range | 99.997% (Metal basis) |
| Beilstein | 3730605 |
| Solubility Information | Soluble in water. |
| IUPAC Name | magnesium(2+) diacetate tetrahydrate |
| Formula Weight | 214.45 (142.39anhydrous) |
| Odor | Odorless |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.