missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate Tetrahydrate, Fisher BioReagents

Product Code. 10521255
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10521255 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10521255 Supplier Fisher BioReagents Supplier No. BP215500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol

Poly Bottle; 500g

Magnesium Acetate is suitable for use in the preparation of buffer solutions and is required as a stabilizer for all stages of bacteriophage purification.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Acetate Tetrahydrate
Melting Point 80°C
CAS 16674-78-5
Color White
pH 8 to 9
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99.0 %
Molecular Formula C4H14MgO8
Linear Formula (CH3COO)2Mg·4H2O
MDL Number MFCD00149214
Merck Index 15, 5718
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
IUPAC Name magnesium(2+) diacetate tetrahydrate
Molecular Weight (g/mol) 214.45
PubChem CID 134717
Formula Weight 214.46
Percent Purity ≥99.0%
Identification Pass Test
Packaging Poly Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.