missing translation for 'onlineSavingsMsg'
Learn More

Melford Magnesium Acetate Tetrahydrate

Product Code. 30018482
Change view
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018482 500 g 500g
30018483 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018482 Supplier Melford Supplier No. M24100500.0
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • CAS Number: 16674-78-5
  • Molecular Formula: (CH3COO)2Mg 4H2O
  • Molecular Weight: 214.45

Specifications

Chemical Name or Material Magnesium Acetate Tetrahydrate
CAS 16674-78-5
Quantity 500 g
Purity Grade Notes Molecular Biology Grade
Molecular Formula C4H6MgO4-4H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.