missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Acetate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 500 g |
| IUPAC Name | magnesium;diacetate;tetrahydrate |
| Formula Weight | 214.45 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.45kg |
| Chemical Name or Material | Magnesium Acetate, 4-Hydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction