missing translation for 'onlineSavingsMsg'
Learn More

Magnesium 2,4-pentanedionate hydrate, typically 98+% (metals basis)

Product Code. 11357367
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
500 g
Unit Size:
10g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357367 250 g 250g
11430290 10 g 10g
11377367 50 g 50g
11367367 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11357367 Supplier Thermo Scientific Alfa Aesar Supplier No. 012532.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67032-77-3 | C10H14MgO4 | 222.523 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67032-77-3
Molecular Formula C10H14MgO4
Molecular Weight (g/mol) 222.523
MDL Number MFCD00150168
InChI Key AKTIAGQCYPCKFX-SYWGCQIGSA-L
Synonym 4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one
PubChem CID 25113476
IUPAC Name magnesium;(E)-4-oxopent-2-en-2-olate
SMILES CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2]

Specifications

Melting Point 260°C (decomposition)
Color White
Quantity 250 g
Formula Weight 222.52 (Anhydrous)
Percent Purity ≥98% (typically)
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Magnesium 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.