missing translation for 'onlineSavingsMsg'
Learn More

Magnamycin B (>80%), TRC

Product Code. 30357417
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357417 1 mg 1mg
30352324 10 mg 10mg
30356747 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357417 Supplier Toronto Research Chemicals Supplier No. TRCC1766851MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21238-30-2
Molecular Formula C42H67NO15
Molecular Weight (g/mol) 825.98
Synonym Carbomycin B, Oxacyclohexadecane Leucomycin V Deriv., 9-Deoxy-9-oxo-Leucomycin V 3-Acetate 4B-(3-Methylbutanoate)
IUPAC Name [(2S,3S,4R,6S)-6-[(2R,3S,4R,5R,6S)-6-[[(4R,5S,6S,7R,9R,11E,13E,16R)-4-acetyloxy-5-methoxy-9,16-dimethyl-2,10-dioxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-6-yl]oxy]-4-(dimethylamino)-5-hydroxy-2-methyloxan-3-yl]oxy-4-hydroxy-2,4-dimethyloxan-3-yl] 3-methylbutanoate
SMILES CO[C@H]1[C@@H](CC(=O)O[C@H](C)C\C=C\C=C\C(=O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)OC(=O)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C42H67NO15/c1-23(2)19-32(47)56-40-27(6)53-34(22-42(40,8)50)57-37-26(5)54-41(36(49)35(37)43(9)10)58-38-29(17-18-44)20-24(3)30(46)16-14-12-13-15-25(4)52-33(48)21-31(39(38)51-11)55-28(7)45/h12-14,16,18,23-27,29,31,34-41,49-50H,15,17,19-22H2,1-11H3/b13-12+,16-14+/t24-,25-,26-,27+,29+,31-,34+,35-,36-,37-,38+,39+,40+,41+,42-/m1/s1
Formula Weight 825.4511
Chemical Name or Material Magnamycin B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.