missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagnaBind™ Magnetic Beads and Magnets

Product Code. p-4531519 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Microcentrifuge Tube (1.5 mL)
Microcentrifuge Tubes (6 x 1.5 mL)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10229853

Brand: Thermo Scientific™ 21357

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enable benchtop magnetic affinity purification with these ligand-coated (streptavidin, Protein A and G, GAM IgG, GAR IgG) magnetic particles and magnet separators.

Thermo Scientific™ MagnaBind Magnetic Beads are available with streptavidin, Protein A, Protein G, GAM IgG, GAR IgG and other ligands for use in small-scale benchtop magnetic affinity protocols.

MagnaBind Magnetic Beads enable small-scale affinity purification and cell-separation without columns or centrifugation. The beads are silanized iron oxide magnetic particles activated with various affinity ligands. After a binding step in an affinity purification procedure, the magnetic particles are easily and rapidly collected by placing the microcentrifuge tube or reaction vessel next to an appropriate rare-earth magnet. MagnaBind Beads respond rapidly to MagnaBind Magnets. The particles can be easily dispersed and regathered multiple times (i.e., they will not irreversibly aggregate) because they do not have any magnetic memory.

MagnaBind Beads are available pre-coated with Protein A, Protein G, streptavidin, anti-mouse or anti-rabbit antibodies. Also available are beads activated with either amine or carboxyl groups as targets for customized covalent immobilization other proteins or ligands.

Highlights:

  • Available pre-coated with popular affinity ligands or derivatized for covalent attachment of proteins and other specific ligands
  • Beads do not irreversibly aggregate because they have no magnetic memory; collect and disperse the beads multiple times if needed
  • Most separations require a short five- to 10-minute benchtop procedure

Properties of underivatized MagnaBind Beads:

  • Composition: silanized iron oxide
  • Magnetization: 25 to 35 EMU/g
  • Type of Magnetization: superparamagnetic (no magnetic memory)
  • Surface Area: greater than 100m2/g
  • Bead Size: 1 to 4μm diameter
  • Settling Rate: 4% in 30 minutes
  • Effective Density: 2.5g/mL
  • Number of Beads: 1 x 108 beads per milliliter
  • pH Stability: aqueous solution, above pH 4.0
  • Concentration: 5mg/mL

Requires:

To remove MagnaBind Beads from suspension, an external magnetic field is used; see the ordering table for a selection of magnets.

Recommended for:

Cell sorting using either positive or negative selection; Protein purification or immunoassays using either direct or indirect methods; Immunoprecipiation

TRUSTED_SUSTAINABILITY

Specifications

Formulation Plastic holder containing strong, rare-earth magnet
For Use With (Equipment) Microcentrifuge Tube (1.5 mL)
Product Line MagnaBind
Quantity 1 Magnet
Type Magnetic Separator
Sufficient For Collecting magnetic particles in 1 microcentrifuge tube at a time
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt