Learn More
Description
Thermo Scientific™ MagnaBind™ Beads provide a convenient method for magnetic separation of antibodies, antigens, lectins, enzymes, nucleic acids and cells using affinity binding. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.
MagnaBind Goat Anti-Mouse IgG Beads can be used to conveniently separate cells of interest from a cell mixture. The monoclonal or polyclonal antibody to the cell surface antigen is pre-incubated with the appropriate magnetic bead that is then incubated with the cell suspension.
Features of MagnaBind Goat Anti-Mouse IgG Beads:
- Composition: Silanized iron oxide
- Magnetization: 25-35EMU/g
- Type of Magnetization: Superparamagnetic (no magnetic memory)
- Surface Area: >100m2/g
- Bead Size: 1-4μm diameter
- Settling Rate: 4% in 30 minutes
- Effective Density: 2.5g/mL
- Number of Beads: 1 x 108 beads/mg
- pH Stability: Aqueous solution, above pH 4.0
- Concentration: ∼1 mg/mL
Applications:
- Cell sorting using either positive or negative selection
- Protein purification or immunoassays using either direct or indirect methods
Specifications
Specifications
| Concentration | Slurry: ∼1 mg/mL |
| Content And Storage | Upon receipt store products at 4°C. Do not freeze products. Products are shipped at ambient temperature. |
| Description | MagnaBind Goat Anti-Mouse IgG Beads |
| Color | Brown |
| For Use With (Application) | Affinity Purification |
| Product Type | Antibody Coated Bead |
| Ligand Type | Antibody |
| Product Line | MagnaBind |
| Quantity | 50 mL |
| Volume (Metric) | 50 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.