missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ Viral/Pathogen Proteinase K
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Description
This Proteinase K is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more Proteinase K than is provided in the kit.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Sample Type | Liquid Samples (e.g. Serum), Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast |
| For Use With (Application) | Next-Generation Sequencing (NGS) |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| Product Line | MagMAX |
| Product Type | Viral/Pathogen Proteinase K |
| Quantity | 10 mL |
| Shipping Condition | Room Temperature |
| Target | Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction