missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen Nucleic Acid Isolation Kit, for HIV-1 dried blood spots (DBS)

Product Code. 17856501 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17856501 100 reactions Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17856501 Supplier Applied Biosystems™ Supplier No. A53770
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems™ MagMAX™ Viral/Pathogen Nucleic Acid Isolation Kit for HIV-1 dried blood spots is designed to recover viral RNA from dried blood spots (DBS) and plasma for HIV-1 drug resistance testing.

  • Kit is designed to recover viral RNA from dried blood spots (DBS) and plasma for HIV-1 drug resistance testing
  • MagMAX magnetic-bead technology ensures reproducible recovery of high-quality HIV-1 viral RNA compatible with downstream applications such as RT-PCR and Sanger sequencing
  • Automation: Workflows are designed for the KingFisher Flex and KingFisher Duo Prime systems
  • Sensitivity: Extracts amplifiable viral RNA from DBS and plasma samples
    • DBS: >2000 copies/mL
    • Plasma: >1000 copies/mL
  • Duration: Test takes less than 60 minutes: ∼30 min DBS lysis, ∼30 min viral RNA extraction

Benefits of MagMAX magnetic bead–based purification:

  • Beads bind the nucleic acid more efficiently than glass-fiber filters, resulting in higher and more consistent yields
  • Since filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples
  • This clogging issue is of particular concern with protein-rich, large-volume samples such as whole blood or plasma that are commonly used for viral testing
TRUSTED_SUSTAINABILITY

Specifications

Bead Type Magnetic
Content And Storage • 60 mL Dried Blood Spots Lysis Solution
• 55 mL Binding Solution
• 100 mL Wash Buffer
• 10 mL Elution Solution
• 1 mL Proteinase K
• 2 mL Total Nucleic Acid Binding Beads

Detection Method HIV-1 viral RNA compatible with downstream applications such as RT-PCR and Sanger sequencing
Sample Type Whole Blood, Plasma
For Use With (Equipment) KingFisher Duo Prime, KingFisher Flex, Magnetic Stand
No. of Reactions 100 reactions
Quantity 100 reactions

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.