missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen Binding Beads

Product Code. 16366582
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
1000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16366582 20 mL 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16366582 Supplier Applied Biosystems™ Supplier No. A42362
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

These binding beads are included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.

These binding beads are included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42352, A42356, A42357, and A42358). They are made available separately for applications that require more binding beads than are provided in the kit.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.

High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Product Type Viral/Pathogen Binding Beads
Quantity 20 mL
Shipping Condition Room Temperature
For Use With (Application) Next-Generation Sequencing (NGS), Real Time PCR (qPCR), RT-PCR
Purification Target Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids
Sample Type Liquid Samples (e.g. Serum), Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.