missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied BioSystems™ MagMAX™ Saliva gDNA Lysis/Binding Solution
Shop All Applied Biosystems Products
Click to view available options
Quantity:
550 mL
Unit Size:
Each
Description
Includes
MagMAX Saliva gDNA Lysis/Binding Solution
This Lysis/Binding solution is a component of the MagMAX Saliva gDNA Isolation Kit. It is offered here as a stand-alone reagent for applications that require more Lysis/Binding solution than is provided in the kit.
Specifications
Specifications
| For Use With (Application) | Next-Generation Sequencing, PCR |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| Product Line | MagMAX |
| Product Type | Saliva gDNA Lysis/Binding Solution |
| Quantity | 550 mL |
| Shipping Condition | Room Temperature |
| Target | Genomic DNA |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction