missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ Microbiome Bead Plate
Click to view available options
Quantity:
1 plate
Unit Size:
Each
Description
This bead plate is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42357). It is made available separately for applications that require additional plates.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Sample Type | Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| For Use With (Application) | Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| Product Line | MagMAX™ |
| Quantity | 1 plate |
| Shipping Condition | Room Temperature |
| Target | Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction