missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ HMW DNA Kit and Booster

Product Code. 31034278 Shop All Applied Biosystems Products
Change view
Click to view available options
Format:
Kit
Reagent
No. of Reactions:
100 Preps
1000 Preps
Up to 100 Preps
Sample Type:
Fresh or Frozen Blood
Fresh or Frozen Blood, Tissue, Cells
Tissue, Cells
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format No. of Reactions Sample Type
31034278 Reagent 1000 Preps Fresh or Frozen Blood, Tissue, Cells
31034358 Reagent 100 Preps Fresh or Frozen Blood, Tissue, Cells
31034703 Kit Up to 100 Preps Fresh or Frozen Blood
31034615 Kit Up to 100 Preps Tissue, Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31034278 Supplier Applied Biosystems™ Supplier No. A36625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximize the full potential of your long-read sequencing applications with our MagMAX HMW DNA Kit. Designed for reproducibility and efficiency, it provides high-integrity DNA with large intact fragments from diverse samples. Suitable for both manual and automated protocols, it meets the varied needs of your laboratory with minimal processing time.

The MagMAX HMW DNA Kit is a magnetic bead-based nucleic acid purification kit for isolation of high-quality, high molecular weight (HMW) DNA from whole blood, cells, and tissue. This versatile kit is optimized for both automated workflows with KingFisher purification systems and manual isolation processes, providing flexibility and scalability. It is specifically designed to obtain intact DNA fragments larger than 100 kb, making it excellent for long-read sequencing and various downstream molecular applications.

The MagMAX HMW DNA Kit is for use with fresh and frozen whole blood, while the MagMAX HMW DNA Kit with Cell and Tissue Extraction Buffer is also compatible with cell and tissue sample types. Already have MagMAX DNA Multi-Sample Ultra 2.0 and/or MagMAX Cell and Tissue Extraction Buffer? No problem, we offer MagMAX HMW DNA Booster separately in the 100 prep size found in the kit or a 1000 prep size for higher throughput.

Features of the MagMAX HMW DNA Kit include:

  • Reproducibility and efficiency—consistently yields intact DNA fragments exceeding 100 kb from a range of sample types
  • Versatile solution—flexible workflows for low- to high-throughput sample processing with manual or automated protocols
  • Fast and optimized workflow—eliminates the need for post-processing steps or overnight elution
  • Innovate your research—optimal performance with long-read systems such as Oxford Nanopore Technologies and PacBio systems
  • Automation compatibility
  • Available as stand-alone products—reagents available in larger volume bottles sold separately to meet your scalability needs

Versatility for downstream applications

Start your workflow out right with the MagMAX HMW DNA Kit, designed to meet the high demand for high-quality nucleic acid isolation, a vital component for highly sensitive downstream applications. The MagMAX HMW DNA Kit has optimized workflows available with the KingFisher purification systems, enabling a semi-automated workflow with a total turnaround time of less than two hours. This versatile kit is also optimized for lower throughput manual workflows, making it an excellent choice for laboratories of all sizes and throughput needs. Experience the perfect blend of efficiency, flexibility, and reliability with the MagMAX HMW DNA Kit for your downstream molecular applications, such as long-read sequencing.

Isolate HMW DNA from multiple sample types with a single sample prep kit

The MagMAX HMW DNA Kit leverages the innovative MagMAX HMW DNA Booster to enhance the elongation of fragment size. It is designed to isolate large fragment gDNA (100–300 kb) from various sample types, including 200 μL of fresh or frozen whole blood, one million cultured cells, and 2–8 mg tissue samples. Dynabeads magnetic beads with optimized surface chemistry and high specific surface area enable efficient kinetics and high sensitivity in capturing nucleic acids. The robust chemistry and workflow design minimizes impurities, such as proteins and detergents, that can interfere with downstream molecular analysis. Finally, the HMW genomic DNA is released from the beads using a low quantity TE elution solution.

The MagMAX HMW DNA Kit leverages the MagMAX DNA Multi-sample Ultra 2.0 Kit chemistry. This kit is specifically designed for the extraction of high molecular weight DNA from fresh and frozen whole blood samples. For cell and tissue samples, the MagMAX HMW DNA Kit with Cell and Tissue DNA Extraction Buffer provides an optimized solution, delivering reliable and efficient nucleic acid isolation. Existing users of the MagMAX DNA Multi-sample Ultra 2.0 Kit (A36570) can enhance their workflow by incorporating the MagMAX HMW DNA Booster (A36624, A36625) for whole blood, along with the MagMAX Cell and Tissue DNA Extraction Buffer (A45469, A45470) for cells and tissue. This combination enables the isolation of longer DNA fragments, thus broadening the scope of your genomic research.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1.0 mL MagMAX HMW DNA Booster, 1000 preps
Format Reagent
Isolation Technology Magnetic Bead
Sample Type Fresh or Frozen Blood, Tissue, Cells
For Use With (Application) Long-read sequencing, and other molecular applications
No. of Reactions 1000 Preps
Product Line MagMAX
Product Type DNA Booster
Quantity 1 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.