missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit

Product Code. p-9056014 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
2500 Preps
500 Preps
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15474773

Brand: Applied Biosystems™ A25597

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue.

The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue. gDNA purified with the kit is ideal for qPCR applications on sensitive, scalable platforms such as the QuantStudio™ 12K Flex System with OpenArray™ Block (with AccuFill™ System). The magnetic bead–based purification format allows you to easily scale from processing 12 to 500 samples a day, making it the ideal choice for pharmacogenomics (PGx) studies.

Features of the MagMAX™ DNA Multi-Sample Ultra Kit include:
• Streamlined protocols for numerous non-invasive biological samples
• Optimized to provide DNA yields suitable for OpenArray™ and Ion AmpliSeq™ applications
• Packaged for high-throughput needs

One Kit for a Diverse Set of Sample Types
Why purchase multiple purification kits to isolate gDNA from different sample types? The MagMAX™ DNA Multi-Sample Ultra Kit eliminates the headache of ordering these extra kits because it has been optimized for use with a variety of biological samples. The kit is designed for high-throughput isolation of gDNA from 1 buccal swab, 50 to 350 μL of whole blood, up to 400 μL urine (without sample concentration), up to 150 μL saliva, 1-5 mL mouth rinse, blood spot cards, and mammalian tissue samples (up to 10 mg). The protocols and magnetic bead format make the kit fully amenable to automation.

*Please note that the Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processor with a 24 Deep-well Head is recommended for sample volumes greater than 50 μL.

Benefits of MagMAX™ Magnetic Bead–based Purification
Magnetic beads offer many benefits compared to other technologies for isolating gDNA. Beads bind gDNA more efficiently than glass fiber filters, resulting in higher and more consistent gDNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples.

Accessory Products
Proteinase K (Cat. No. A25561), DNA Binding Beads (Cat. No. A25562), Wash Solution 1 Concentrate (Cat. No. AM8504), and Wash Solution 2 Concentrate (Cat. No. AM8640) are components of the kit that may also be purchased separately.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • DNA Binding Beads, 1 × 8 mL (store at 2–8°C, DO NOT FREEZE)
• DNA Elution Buffer 1, 1 x 25 mL (store at room temperature)
• DNA Elution Buffer 2, 1 x 25 mL (store at room temperature)
• Wash Solution 1 Concentrate, 1 x 80 mL* (store at room temperature)
• Wash Solution 2 Concentrate, 1 x 162 mL* (store at room temperature)
• Multi-Sample DNA Lysis Buffer, 1 x 100 mL (store at room temperature)
• Nuclease-Free Water, 1 x 100 mL (store at room temperature)
• PK Buffer, 1 x 96 mL (store at room temperature)
• Proteinase K, 1 x 4 mL (store at -5 to -30°C)
• Ribonuclease A (RNase A), 1 x 1.25 mL (store at -5 to -30°C)
*after addition of alcohol
Format Kit
Isolation Technology Magnetic Bead
Sample Type PCR Products
For Use With (Application) Real-Time PCR, Sequencing
No. of Reactions 500 Preps
Product Line MagMAX
Product Type Kit, 500 preps
Quantity 500 Preps

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.