missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry
Shop All Applied Biosystems Products
Click to view available options
Quantity:
100 tests
Unit Size:
Pack of 100
Description
MagMAX™ CORE Glass Microbeads are a supplement to the MagMAX™ CORE Nucleic Acid Purification Kit and MagMAX™ CORE Mechanical Lysis Module. The beads aid in the homogenization of completely desiccated feces such as environmental samples or fecal pellets.
The Mechanical Lysis Module is also available separately.
Specifications
Specifications
| Content And Storage | • 1 x 50 mL MagMAX CORE Glass Microbeads • 1 x 50 mL MagMAX CORE Lysis Solution • 1 x 45 mL MagMAX CORE Binding Solution • 1 x 60 mL MagMAX CORE Wash Solution 1 • 1 x 60 mL MagMAX CORE Wash Solution 2 • 1 x 12 mL MagMAX CORE Elution Buffer • 1 x 2.2 mL MagMAX CORE Magnetic Beads • 1 x 1.25 mL MagMAX CORE Proteinase K • 1 x 45 mL MagMAX CORE Clarifying Solution • 100 MagMAX CORE Bead Beating Tubes (2 mL) |
| Sample Type | Bacteria, Blood, Buccal Samples, Cells, DNA, RNA, Tissue, Virus |
| For Use With (Application) | Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| High-throughput Compatibility | Automated Protocols, Manual Protocols, Multiplexing |
| Product Line | MagMAX™ |
| Quantity | 100 tests |
| Shipping Condition | Room Temperature |
| Target | Total RNA, Pathogen DNA⁄RNA, Genomic DNA, Viral DNA⁄RNA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction