missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ CORE Lysis Solution

Product Code. 30057785 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30057785 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30057785 Supplier Applied Biosystems™ Supplier No. A32837

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This MagMAX™ Lysis Solution is made available as a supplement to that included in the MagMAX™ CORE Nucleic Acid Purification Kit.

This proprietary buffer solution is used to break open cells in preparation for downstream molecular detection applications such as real time PCR, RT-PCR, and sequencing applications.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 50 mL MagMAX CORE Lysis Solution
Store at room temperature.
Sample Type Bacteria, Blood, DNA, Liquid Samples (e.g. Serum), RNA, Tissue, Virus
For Use With (Application) Real-Time PCR, Sequencing
High-throughput Compatibility High-throughput Compatible
Product Line MagMAX
Quantity 50 mL
Shipping Condition Room Temperature
Target Genomic DNA, Pathogen DNA/RNA, Total Nucleic Acids, Viral DNA/RNA

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.