missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Cell and Tissue DNA Extraction Buffer

Product Code. 17165772
Change view
Click to view available options
No. of Reactions:
100 Preps
1000 Preps
Unit Size:
1000 preps
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
17165772 1000 Preps 1000 preps
16584810 100 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17165772 Supplier Applied Biosystems™ Supplier No. A45470
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 600 mL Cell and Tissue DNA Extraction Buffer
Sample Type Cells, Tissue
For Use With (Application) Next-Generation Sequencing, PCR, Real-Time PCR
High-throughput Compatibility Automated Protocols, High-throughput Compatible, Manual Protocols
No. of Reactions 1000 Preps
Product Line MagMAX
Product Type Cell and Tissue DNA Extraction Buffer
Quantity 1000 Preps
Shipping Condition Room Temperature
Target Genomic DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.