missing translation for 'onlineSavingsMsg'
Learn More

Magmas Antibody, Novus Biologicals™

Product Code. 18338461 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338461 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338461 Supplier Novus Biologicals Supplier No. NBP318383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Magmas Polyclonal antibody specifically detects Magmas in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Magmas
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Immunoprecipitation 8 - 15 ug/mg lysate
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias MAGMAS, mitochondria associated protein involved in granulocyte macrophage colonystimulating factor signal transduction, Mitochondria-associated granulocyte macrophage CSF-signaling molecule, mitochondrial import inner membrane translocase subunit TIM16, presequence translocase-associated motor 16 homolog (S. cerevisiae), Presequence translocated-associated motor subunit PAM16, TIM16, TIMM16magmas-like protein
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 75 and 125..
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51025
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.