missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady Magicard™ 300/600 - Printhead Assembly-MK2
Click to view available options
Unit Size:
Each
Description
- Easy to install, no tools required.
- High quality printhead.
- Fits both the Magicard 300 and Magicard 600.
Specifications
Specifications
| Product Type | Printhead |
| For Use With (Equipment) | Magicard 300, Magicard 600 |
| Description | Printhead assembly for Magicard 300/600 printer |
| Format | Printhead |
| Includes | 1 Printhead Assembly |
| For Use With (Application) | Healthcare and Hospital labelling;General identification;Facility identification;Security identification |
| Quantity | 1 Ea. |
| Alternate Part Number | FG/3652-0160-2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction