missing translation for 'onlineSavingsMsg'
Learn More

MAGI3 Sheep anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18380103
Change view
Click to view available options
Quantity:
25 μg
250 μg
Unit Size:
250µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380103 250 μg 250µg
18386222 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18380103 Supplier Bio-Techne Supplier No. NBP311883250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

MAGI3 Polyclonal antibody specifically detects MAGI3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGI3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 10 ug/ml
Formulation PBS
Gene Alias dJ730K3.2, MAGI-3, membrane associated guanylate kinase, WW and PDZ domain containing 3, membrane-associated guanylate kinase inverted 3, membrane-associated guanylate kinase-related 3, WW and PDZ domain-containing protein 3
Host Species Sheep
Immunogen Recombinant protein corresponding to the guanylate kinase-WW domain of MAGI3. (Uniprot: Q9H2V6)
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260425
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.