missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGI2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25189-1-AP-20UL
This item is not returnable.
View return policy
Description
The MAGI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MAGI2. This antibody recognizes human, mouse, rat antigen. The MAGI2 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| MAGI2 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9863 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 800 μg/mL | |
| ACVRIP1; AIP1; ARIP1; MAGI-2; SSCAM | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 140 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering