missing translation for 'onlineSavingsMsg'
Learn More

MAGI2 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30737733 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737733 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737733 Supplier Novus Biologicals Supplier No. AF7117E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

MAGI2 Polyclonal antibody specifically detects MAGI2 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGI2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias ACVRINP1, ACVRIP1, AIP1AIP-1, ARIP1, atrophin 1 interacting protein 1, atrophin-1 interacting protein A, Atrophin-1-interacting protein 1, Atrophin-1-interacting protein A, KIAA0705activin receptor interacting protein 1, MAGI-2SSCAM, membrane associated guanylate kinase, WW and PDZ domain containing 2, Membrane-associated guanylate kinase inverted 2, membrane-associated guanylate kinase, WW and PDZ domain-containing protein 2
Host Species Goat
Immunogen E. coli-derived recombinant human MAGI2, Ser2-Arg130, Accession # Q86UL8
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9863
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.