missing translation for 'onlineSavingsMsg'
Learn More

MAGI1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18340266 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340266 25 μg 25µL
18314054 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18340266 Supplier Novus Biologicals Supplier No. NBP31788525UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAGI1 Polyclonal antibody specifically detects MAGI1 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen MAGI1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias AIP-3, AIP3WW and PDZ domain-containing protein 1, membrane associated guanylate kinase, WW and PDZ domain containing 1, TNRC19BAP-1, WWP3WW domain-containing protein 3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: ITNKSHSDIVNLIKEAGNTVTLRIIPGDESSNATLLTNAEKIATITTTHTPSQQGTQETRNTTKPKQESQFEFKAPQATQEQDF
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9223
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.