missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55048-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to MAGI1. The antibody can recognize all isoforms of MAGI1.
The protein encoded by this gene is a member of the membrane-associated guanylate kinase homologue (MAGUK) family. MAGUK proteins participate in the assembly of multiprotein complexes on the inner surface of the plasma membrane at regions of cell-cell contact. The product of this gene may play a role as scaffolding protein at cell-cell junctions. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
| MAGI1 | |
| Polyclonal | |
| Unconjugated | |
| MAGI1 | |
| AIP 3, AIP3, BAI1 associated protein 1, BAIAP1, BAP 1, BAP1, MAGI 1, MAGI1, TNRC19, WW domain containing protein 3, WWP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14924, 500261, 9223 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.74 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4L1J4, Q6RHR9, Q96QZ7 | |
| Magi1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction