missing translation for 'onlineSavingsMsg'
Learn More

MAGI1 Antibody (7B4), Novus Biologicals™

Product Code. 18377118 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377118 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377118 Supplier Novus Biologicals Supplier No. H00009223M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAGI1 Monoclonal antibody specifically detects MAGI1 in Human, Rat, Canine, Hamster samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGI1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 7B4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100 to 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 to 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004733
Gene Alias AIP-3, AIP3WW and PDZ domain-containing protein 1, membrane associated guanylate kinase, WW and PDZ domain containing 1, TNRC19BAP-1, WWP3WW domain-containing protein 3
Host Species Mouse
Immunogen MAGI1 (NP_004733, 761 a.a. ~ 859 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SHSTQVLPEFPPAEAQAPDQTDSSGQKKPDPFKIWAQSRSMYENRPMSPSPASGLSKGEREREINSTNFGECPIPDYQEQDIFLWRKETGFGFRILGGN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9223
Target Species Human, Rat, Canine, Hamster
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.