missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific MagFuge™ Silicone Lid Mat

MagFuge™ Silicone Lid Mat

Brand:  Heathrow Scientific 120682

Additional Details : Weight : 0.02000kg

Product Code. 16320896

  • £21.60 / Each
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Silicone Lid Mat
Specifications

Specifications

Silicone
12.80 cm
Stirrer
5.04 in.
White
Bag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers