missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599777
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAGH1.
Melanoma-associated antigen (MAGE) are completely silent in normal tissues, with the exception of male germ cells, and, for some of them, placenta. These antigens ought to be strictly tumor specific, expressed in tumor cells of various histological types. Because of their specific expression on tumor cells, these antigens are of particular interest for antitumor immunotherapy. Genes of the MAGE family direct the expression of tumor antigens that are recognized on a human melanoma by autologous cytolytic T lymphocytes. Though the function of MAGEH1 is not fully elucidated, it may play a role in embryonal development and tumor transformation or aspects of tumor progression.
Specifications
| MAGEH1 | |
| Polyclonal | |
| Unconjugated | |
| MAGEH1 | |
| 1-Apr; 2010107K23Rik; Apoptosis-related protein 1; APR1; APR-1; MAGE family member H1; MAGEH; Mageh1; MAGE-H1 antigen; melanoma antigen family H, 1; melanoma antigen family H1; melanoma antigen, family H, 1; melanoma-associated antigen H1; Restin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 28986, 367767, 75625 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5PPP4, Q9H213, Q9NWG9 | |
| MAGEH1 | |
| A synthesized peptide derived from human MAGEH1(Accession Q9H213), corresponding to amino acid residues M1-S42. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction