missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550701
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MAGEF1 protein.
Melanoma-associated antigen (MAGE) are completely silent in normal tissues, with the exception of male germ cells, and, for some of them, placenta. These antigens ought to be strictly tumor specific, expressed in tumor cells of various histological types. Because of their specific expression on tumor cells, these antigens are of particular interest for antitumor immunotherapy. Genes of the MAGE family direct the expression of tumor antigens that are recognized on a human melanoma by autologous cytolytic T lymphocytes. Though the function of MAGE is unknown, may play a role in embryonal development and tumor transformation or aspects of tumor progression.
Specifications
| MAGEF1 | |
| Polyclonal | |
| Unconjugated | |
| MAGEF1 | |
| 6430590I03Rik; MAGE family member F1; MAGEF1; MAGE-F1; MAGE-F1 antigen; MAGF1; melanoma antigen family F, 1; melanoma antigen family F1; melanoma-associated antigen F1; MGC19617 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64110 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9HAY2 | |
| MAGEF1 | |
| Synthetic peptide corresponding to residues near the N terminal of human melanoma antigen family F, 1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction