missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGED1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565365
This item is not returnable.
View return policy
Description
Immunogen sequence: ARPKSAFKVQN ATTKGPNGVY DFSQAHNAKD VPNTQPKAAF KSQNATPKGP NAAYDFSQAA TTGELAANKS EMAFKAQNAT TKVGPNATYN FSQSLNANDL ANSRPKTPFK AWNDTTKAPT ADTQTQNVNQ AKMATSQADI ETDPGISEP Highest antigen sequence identity to the following orthologs - mouse 67%, rat 68%.
This gene is a member of the melanoma antigen gene (MAGE) family. Most of the genes of this family encode tumor specific antigens that are not expressed in normal adult tissues except testis. Although the protein encoded by this gene shares strong homology with members of the MAGE family, it is expressed in almost all normal adult tissues. This gene has been demonstrated to be involved in the p75 neurotrophin receptor mediated programmed cell death pathway. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| MAGED1 | |
| Polyclonal | |
| Unconjugated | |
| MAGED1 | |
| 11B6; 2810433C11Rik; 5430405L04Rik; BCG-1; Dlixin; Dlxin1; DLXIN-1; DXBwg1492e; JCL-1; MAGE family member D1; MAGE tumor antigen CCF; MAGED1; MAGE-D1; MAGE-D1 antigen; Maged2; melanoma antigen family D, 1; melanoma antigen family D1; melanoma antigen, family D, 1; melanoma-associated antigen D1; mFLJ00163; neurotrophin receptor-interacting MAGE homolog; NRAGE; PP2250; PRO2292; Sertoli cell necdin-related gene protein 1; SNERG-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9500 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5V3 | |
| MAGED1 | |
| Recombinant Human MAGED1. Recombinant protein control fragment (Product #RP-104776). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction