missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103283
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAGEC1.
MAGEC1 is a member of the melanoma antigen gene (MAGE) family. The proteins of this family are tumor-specific antigens that can be recognized by autologous cytolytic T lymphocytes. This protein contains a large number of unique short repetitive sequences in front of the MAGE-homologous sequence, thus is about 800 residues longer than the other MAGE proteins. This gene is expressed in a significant proportion of tumors of various histological types, whereas it is silent in normal tissues, except testis. The function of this gene is unknown.
Specifications
| MAGEC1 | |
| Polyclonal | |
| Unconjugated | |
| MAGEC1 | |
| Cancer/testis antigen 7.1; cancer/testis antigen family 7, member 1; CT7; CT7.1; MAGE family member C1; MAGEC1; MAGE-C1 antigen; melanoma antigen family C, 1; melanoma antigen family C1; melanoma-associated antigen C1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9947 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60732 | |
| MAGEC1 | |
| A synthesized peptide derived from human MAGEC1(Accession O60732), corresponding to amino acid residues P1100-E1142. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction