missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAGEB4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA556478

Product Code. 15724391

  • £483.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: KVGQPTAAEK EESPSSSSSV LRDTASSSLA FGIPQEPQRE PPTTSAAAAM SCTGSDKGDE SQDEENASSS QASTSTERSL KDSLTRKTKM LVQFLLYKYK MKEPTTKAEM LKIISKKYKE HFPE Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 36%.

This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEB genes are clustered on chromosome Xp22-p21. This gene sequence ends in the first intron of MAGEB1, another family member. This gene is expressed in testis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAGEB4
Polyclonal
Unconjugated
MAGEB4
cancer/testis antigen family 3, member 6; CT3.6; MAGE family member B4; MAGEB4; MAGE-B4 antigen; melanoma antigen family B, 4; melanoma antigen family B4; melanoma-associated antigen B4; MGC33144
Rabbit
Antigen affinity chromatography
RUO
4115
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O15481
MAGEB4
Recombinant protein corresponding to Human MAGEB4. Recombinant protein control fragment (Product #RP-92272).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.