missing translation for 'onlineSavingsMsg'
Learn More

MAGEB4 Antibody (OTI3H2) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30688479 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688479 Supplier Novus Biologicals Supplier No. NBP344915CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAGEB4 Monoclonal antibody specifically detects MAGEB4 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI3H2
Conjugate CoraFluor 1
Formulation PBS
Gene Alias cancer/testis antigen family 3, member 6, CT3.6, MAGE-B4 antigen, melanoma antigen family B, 4, melanoma-associated antigen B4, MGC33144
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-193 of human MAGEB4(NP_002358) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4115
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.