missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoH Antibody (OTI9F4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45048-100ul
This item is not returnable.
View return policy
Description
RhoH Monoclonal antibody specifically detects RhoH in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| RhoH | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9F4 | |
| Western Blot 1:2000 | |
| ras homolog gene family, member H, translocation three four | |
| Human recombinant protein fragment corresponding to amino acids 97-190 of human RHOH(NP_004301) produced in E.coli. | |
| 100 μL | |
| Signal Transduction | |
| 399 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction