missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB3 Antibody (OTI5H8), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72579PCP
This item is not returnable.
View return policy
Description
MAGEB3 Monoclonal antibody specifically detects MAGEB3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MAGEB3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 4114 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5H8 | |
| Western Blot, Immunohistochemistry | |
| CT3.5, MAGE-B3 antigen, melanoma antigen family B, 3, melanoma-associated antigen B3, member 5 | |
| Full length human recombinant protein of human MAGEB3(NP_002356) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction