missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB3 Antibody (OTI5H8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72579FR
This item is not returnable.
View return policy
Description
MAGEB3 Monoclonal specifically detects MAGEB3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MAGEB3 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| MAGEB3 | |
| Full length recombinant protein of human MAGEB3 (NP_002356) produced in E.coli. | |
| 0.1 mL | |
| 4114.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI5H8 | |
| Western Blot, Immunohistochemistry | |
| CT3.5, MAGE-B3 antigen, melanoma antigen family B, 3, melanoma-associated antigen B3, member 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction